Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBFOX1 SIRNA Assays ›
Gene Symbol:
RBFOX1
Gene Name:
RNA binding protein, fox-1 homolog 1
Gene Aliases:
2BP1, A2BP1, FOX-1, FOX1, HRNBP1
Chromosome Location:
Chr.16: 5239749 - 7713343 on Build GRCh38
UniGene ID:
Hs.459842
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29340
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBFOX1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142333.1
NP_001135805.1
9
1592
NM_001142334.1
NP_001135806.1
7
824
NM_001308117.1
NP_001295046.1
9
929
NM_018723.3
NP_061193.2
9
1592
NM_145891.2
NP_665898.1
6
916
NM_145892.2
NP_665899.1
6
916
NM_145893.2
NP_665900.1
6
916
XM_005255386.3
XP_005255443.1
6
842
XM_005255387.3
XP_005255444.1
6
821
XM_005255388.4
XP_005255445.1
6
877
XM_005255390.3
XP_005255447.1
9
1714
XM_005255391.3
XP_005255448.1
8
724
XM_005255394.4
XP_005255451.1
6
821
XM_011522546.2
XP_011520848.1
6
821
XM_011522547.2
XP_011520849.1
6
821
XM_011522548.2
XP_011520850.1
6
821
XM_017023318.1
XP_016878807.1
10
1219
XM_017023319.1
XP_016878808.1
12
1215
XM_017023320.1
XP_016878809.1
9
1623
XM_017023321.1
XP_016878810.1
8
2685
XM_017023322.1
XP_016878811.1
8
2685
XM_017023323.1
XP_016878812.1
6
842
XM_017023324.1
XP_016878813.1
9
1623
XM_017023325.1
XP_016878814.1
7
1221
XM_017023326.1
XP_016878815.1
9
1714
XM_017023327.1
XP_016878816.1
6
842
XM_017023328.1
XP_016878817.1
9
1623
XM_017023329.1
XP_016878818.1
9
1623
XM_017023330.1
XP_016878819.1
8
2162
XM_017023331.1
XP_016878820.1
6
2628
XM_017023332.1
XP_016878821.1
6
842
XM_017023333.1
XP_016878822.1
8
724
XM_017023334.1
XP_016878823.1
6
842
XM_017023335.1
XP_016878824.1
9
1714
XM_017023336.1
XP_016878825.1
8
724
XM_017023337.1
XP_016878826.1
6
842
XM_017023338.1
XP_016878827.1
8
724
XM_017023339.1
XP_016878828.1
6
868
XM_017023340.1
XP_016878829.1
7
631
XM_017023341.1
XP_016878830.1
9
1714
XM_017023342.1
XP_016878831.1
8
724
XM_017023343.1
XP_016878832.1
6
871
GenBank mRNA
AB208826.1
-
1626
AF094849.1
-
6
917
AF107203.1
-
9
1590
AF109106.1
-
6
917
AF229057.1
-
6
919
AF448859.1
-
9
1590
AK001027.1
-
7
824
AK293951.1
-
9
929
AK299664.1
-
6
502
AL713700.1
-
8
1439
BC113691.1
-
6
710
8322743
BC143816.1
-
6
710
KJ534926.1
-
6
664
KJ535000.1
-
6
664
KJ535001.1
-
6
664
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of alternative mRNA splicing, via spliceosome
mRNA processing
nervous system development
RNA splicing
RNA transport
nucleotide binding
RNA binding
mRNA binding
protein binding
protein C-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0