Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017515.4 | NP_059985.2 | 4 | 905 | ||
GenBank mRNA | AK023080.1 | - | 4 | 502 | ||
AK128062.1 | - | 7 | 1074 | |||
AK300582.1 | - | 3 | 340 | |||
AL833969.1 | - | 4 | 508 | |||
BC039195.2 | - | 4 | 645 | 3847013 | ||
BC048302.1 | - | 4 | 511 | 5499821 |