Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017515.4 | NP_059985.2 | 2 | 565 | ||
GenBank mRNA | AK128062.1 | - | 5 | 734 | ||
AL833969.1 | - | 2 | 168 | |||
BC039195.2 | - | 2 | 305 | 3847013 | ||
BC048302.1 | - | 2 | 171 | 5499821 | ||
X99961.1 | - | 2 | 108 |