Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEPROT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198681.1 | NP_001185610.1 | 4 | 403 | ||
NM_017526.4 | NP_059996.1 | 3 | 539 | |||
GenBank mRNA | AK074841.1 | - | 3 | 300 | ||
AK309370.1 | - | 4 | 403 | |||
BC011027.2 | - | 2 | 247 | |||
BC056250.1 | - | 3 | 284 | 4328021 | ||
CB140261.1 | - | 3 | 407 | |||
CR541647.1 | - | 3 | 262 | |||
CR541737.1 | - | 3 | 262 | |||
Y12670.1 | - | 3 | 332 |
Gene Symbol: LEPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX475249.1 | - | 3 | 352 |