Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPDR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242946.1 | NP_001229875.2 | 2 | 710 | ||
NM_001242948.1 | NP_001229877.1 | 3 | 404 | |||
NM_017549.4 | NP_060019.2 | 3 | 919 | |||
GenBank mRNA | AJ250475.2 | - | 3 | 392 | ||
AK290885.1 | - | 3 | 613 | |||
AK309180.1 | - | 3 | 391 | |||
AY027862.2 | - | 3 | 919 | |||
BC000686.2 | - | 3 | 663 | |||
BC018299.1 | - | 3 | 811 | 3461888 | ||
DA938402.1 | - | 4 | 432 | |||
DB167672.1 | - | 3 | 405 | |||
DQ914439.1 | - | 2 | 331 |