Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017559.2 | NP_060029.1 | 3 | 800 | ||
GenBank mRNA | AK225743.1 | - | 3 | 765 | ||
AK313775.1 | - | 3 | 833 | |||
AL133054.1 | - | 3 | 751 | |||
BC024002.2 | - | 3 | 781 | 4821771 |