Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TET2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127208.2 | NP_001120680.1 | 3 | 1270 | ||
NM_017628.4 | NP_060098.3 | 3 | 1187 | |||
XM_005263082.2 | XP_005263139.1 | 2 | 1146 | |||
XM_006714242.3 | XP_006714305.1 | 3 | 1114 | |||
XM_017008319.1 | XP_016863808.1 | 3 | 1114 | |||
GenBank mRNA | AK055149.1 | - | 3 | 1077 | ||
BC150180.1 | - | 2 | 886 | |||
BC150201.1 | - | 2 | 886 | |||
BX640738.1 | - | 3 | 1188 | |||
FM992369.1 | - | 3 | 1643 |