Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317939.1 | NP_001304868.1 | 3 | 418 | ||
NM_001317940.1 | NP_001304869.1 | 3 | 324 | |||
NM_017637.5 | NP_060107.3 | 4 | 544 | |||
XM_011517924.1 | XP_011516226.1 | 3 | 468 | |||
XM_017014816.1 | XP_016870305.1 | 6 | 2854 | |||
XM_017014817.1 | XP_016870306.1 | 5 | 1003 | |||
XM_017014818.1 | XP_016870307.1 | 4 | 918 | |||
XM_017014819.1 | XP_016870308.1 | 5 | 1002 | |||
XM_017014820.1 | XP_016870309.1 | 5 | 1002 | |||
XM_017014821.1 | XP_016870310.1 | 4 | 916 | |||
XM_017014822.1 | XP_016870311.1 | 2 | 464 | |||
XM_017014823.1 | XP_016870312.1 | 2 | 5233 | |||
XM_017014825.1 | XP_016870314.1 | 5 | 1001 | |||
XM_017014826.1 | XP_016870315.1 | 5 | 1001 | |||
XM_017014827.1 | XP_016870316.1 | 5 | 1001 | |||
XM_017014828.1 | XP_016870317.1 | 5 | 1000 | |||
XM_017014829.1 | XP_016870318.1 | 2 | 894 | |||
GenBank mRNA | AK131398.1 | - | 4 | 426 | ||
AK299073.1 | - | 3 | 294 | |||
AY438376.1 | - | 2 | 189 | |||
CR933649.1 | - | 3 | 377 | |||
DA494074.1 | - | 4 | 428 | |||
DB282126.1 | - | 3 | 419 | |||
DQ884938.1 | - | 5 | 534 | |||
DQ884939.1 | - | 5 | 530 | |||
DQ884940.1 | - | 4 | 518 | |||
DQ884941.1 | - | 5 | 507 | |||
DQ884944.1 | - | 2 | 443 |