Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127350.1 | NP_001120822.1 | 3 | 758 | ||
NM_017638.2 | NP_060108.2 | 3 | 814 | |||
XM_005245914.4 | XP_005245971.1 | 3 | 728 | |||
GenBank mRNA | AB107222.1 | - | 3 | 770 | ||
AK000052.1 | - | 3 | 778 | |||
AK296130.1 | - | 3 | 805 | |||
BC002694.2 | - | 3 | 761 | 3607295 |