Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270890.1 | NP_001257819.1 | 16 | 2907 | ||
NM_017645.4 | NP_060115.3 | 16 | 3012 | |||
XM_011517935.2 | XP_011516237.1 | 5 | 1860 | |||
XM_017014830.1 | XP_016870319.1 | 12 | 2163 | |||
GenBank mRNA | AB046794.1 | - | 9 | 1868 | ||
AF533709.1 | - | 16 | 3194 | |||
AF537091.1 | - | 16 | 2974 | |||
AK000067.1 | - | 2 | 1100 | |||
AK022964.1 | - | 13 | 2328 | |||
AK023068.1 | - | 9 | 1931 | |||
AK056458.1 | - | 16 | 2974 | |||
AK225209.1 | - | 2 | 1100 | |||
AK301852.1 | - | 9 | 1767 | |||
AL832495.1 | - | 16 | 2972 | |||
BC010632.1 | - | 2 | 969 | |||
BC065935.1 | - | 14 | 2512 | |||
BC111041.1 | - | 12 | 2288 | 5529130 | ||
BC113981.1 | - | 16 | 2809 | 40082296 | ||
BC114492.1 | - | 16 | 2809 | 40082291 |