Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000735.3 | NP_000726.1 | 3 | 403 | ||
NM_001252383.1 | NP_001239312.1 | 4 | 496 | |||
GenBank mRNA | AK311972.1 | - | 3 | 360 | ||
BC010957.1 | - | 3 | 366 | 4043527 | ||
BC020782.1 | - | 3 | 362 | 4732686 | ||
BC055080.1 | - | 3 | 364 | 4044086 | ||
BG571584.1 | - | 4 | 453 | |||
BG619154.1 | - | 3 | 404 | |||
FJ624078.1 | - | 2 | 189 | |||
FJ624079.1 | - | 2 | 261 | |||
V00518.1 | - | 3 | 311 |