Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000735.3 | NP_000726.1 | 2 | 191 | ||
NM_001252383.1 | NP_001239312.1 | 2 | 191 | |||
GenBank mRNA | AK311972.1 | - | 2 | 148 | ||
BC010957.1 | - | 2 | 154 | 4043527 | ||
BC020782.1 | - | 2 | 150 | 4732686 | ||
BC055080.1 | - | 2 | 152 | 4044086 | ||
BG571584.1 | - | 2 | 148 | |||
BG619154.1 | - | 2 | 192 | |||
CR457427.1 | - | 1 | 49 | |||
FJ624079.1 | - | 1 | 49 | |||
V00518.1 | - | 2 | 99 |