Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATAD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300946.1 | NP_001287875.1 | 3 | 487 | ||
NM_017660.3 | NP_060130.3 | 4 | 793 | |||
XM_011528105.1 | XP_011526407.1 | 4 | 676 | |||
XM_017026901.1 | XP_016882390.1 | 3 | 487 | |||
XM_017026902.1 | XP_016882391.1 | 4 | 586 | |||
XM_017026903.1 | XP_016882392.1 | 4 | 676 | |||
XM_017026904.1 | XP_016882393.1 | 4 | 633 | |||
XM_017026905.1 | XP_016882394.1 | 4 | 646 | |||
XM_017026906.1 | XP_016882395.1 | 4 | 676 | |||
XM_017026907.1 | XP_016882396.1 | 5 | 755 | |||
XM_017026908.1 | XP_016882397.1 | 6 | 1015 | |||
XM_017026909.1 | XP_016882398.1 | 6 | 966 | |||
XM_017026910.1 | XP_016882399.1 | 4 | 575 | |||
XM_017026911.1 | XP_016882400.1 | 4 | 747 | |||
GenBank mRNA | AK125974.1 | - | 6 | 898 | ||
AK296781.1 | - | 2 | 213 | |||
AK315416.1 | - | 3 | 363 | |||
AY186731.1 | - | 3 | 481 | |||
BC011684.1 | - | 4 | 522 | 3953055 |