Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017680.4 | NP_060150.4 | 6 | 1082 | ||
GenBank mRNA | AF316824.1 | - | 5 | 800 | ||
AK000136.1 | - | 4 | 452 | |||
AK172737.1 | - | 4 | 552 | |||
AK308107.1 | - | 6 | 996 | |||
AY029191.1 | - | 6 | 1015 | |||
AY358329.1 | - | 6 | 1015 | |||
BC063114.1 | - | 6 | 1047 | 30404602 |