Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIVM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017693.3 | NP_060163.2 | 3 | 919 | ||
GenBank mRNA | AF411385.1 | - | 3 | 919 | ||
AK000166.1 | - | 1 | 131 | |||
AK225229.1 | - | 1 | 131 | |||
AK290658.1 | - | 3 | 788 | |||
AK304391.1 | - | 2 | 673 | |||
BC020663.1 | - | 1 | 63 | |||
BC051813.2 | - | 1 | 44 | |||
BC075084.2 | - | 1 | 267 | 30915598 | ||
BC075085.2 | - | 1 | 267 | 30915386 |
Gene Symbol: BIVM-ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204425.1 | NP_001191354.1 | 1 | 362 | ||
GenBank mRNA | AB209312.1 | - | 1 | 154 | ||
LM994822.1 | - | 1 | 240 |