Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017694.3 | NP_060164.3 | 7 | 2281 | ||
XM_005246654.2 | XP_005246711.1 | 6 | 2159 | |||
XM_005246656.3 | XP_005246713.1 | 6 | 2159 | |||
XM_011511368.1 | XP_011509670.1 | 8 | 2347 | |||
XM_017004392.1 | XP_016859881.1 | 7 | 2281 | |||
GenBank mRNA | AB472299.1 | - | 5 | 1957 | ||
AK000167.1 | - | 5 | 1059 | |||
AK098085.1 | - | 5 | 1691 | |||
AK127176.1 | - | 7 | 2281 | |||
AK225240.1 | - | 5 | 1059 | |||
AK302647.1 | - | 5 | 559 | |||
AK302753.1 | - | 5 | 582 | |||
BC050537.2 | - | 5 | 1220 | 6204874 |