Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DEF8 SIRNA Assays ›
Gene Symbol:
DEF8
Gene Name:
differentially expressed in FDCP 8 homolog (mouse)
Gene Aliases:
-
Chromosome Location:
Chr.16: 89948697 - 89968060 on Build GRCh38
UniGene ID:
Hs.62771
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29579
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DEF8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242816.1
NP_001229745.1
6
564
NM_001242817.1
NP_001229746.1
5
460
NM_001242818.1
NP_001229747.1
6
594
NM_001242819.1
NP_001229748.1
5
507
NM_001242820.1
NP_001229749.1
7
834
NM_001242821.1
NP_001229750.1
5
497
NM_001242822.1
NP_001229751.1
6
591
NM_017702.3
NP_060172.1
6
594
NM_207514.2
NP_997397.1
6
719
XM_005256318.2
XP_005256375.2
6
570
XM_011523156.2
XP_011521458.1
6
570
XM_011523157.2
XP_011521459.1
6
570
XM_011523158.1
XP_011521460.1
6
570
XM_011523159.2
XP_011521461.1
6
518
XM_011523160.2
XP_011521462.1
6
671
XM_011523161.2
XP_011521463.1
6
652
XM_011523162.2
XP_011521464.1
5
531
XM_017023358.1
XP_016878847.1
6
570
XM_017023359.1
XP_016878848.1
6
570
XM_017023360.1
XP_016878849.1
6
570
XM_017023361.1
XP_016878850.1
6
518
XM_017023362.1
XP_016878851.1
6
518
XM_017023363.1
XP_016878852.1
6
652
XM_017023364.1
XP_016878853.1
6
518
XM_017023365.1
XP_016878854.1
6
671
XM_017023366.1
XP_016878855.1
5
1635
XM_017023367.1
XP_016878856.1
5
531
XM_017023368.1
XP_016878857.1
5
1635
XM_017023369.1
XP_016878858.1
5
531
GenBank mRNA
AK000193.1
-
6
530
AK095051.1
-
6
675
AK096485.1
-
4
155
AK131370.1
-
6
500
AK225539.1
-
6
535
AK295880.1
-
7
796
AK296410.1
-
5
396
AK303510.1
-
5
441
BC015482.2
-
5
517
BC105592.1
-
5
437
BC130482.1
-
5
480
BC144038.1
-
5
480
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
intracellular signal transduction
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0