Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017706.4 | NP_060176.2 | 4 | 608 | ||
XM_005268469.2 | XP_005268526.1 | 4 | 608 | |||
XM_017009600.1 | XP_016865089.1 | 5 | 395 | |||
GenBank mRNA | AK025355.1 | - | 4 | 555 | ||
AK056861.1 | - | 3 | 619 | |||
AK122862.1 | - | 2 | 2043 | |||
AK293417.1 | - | 1 | 95 | |||
BC002482.2 | - | 3 | 314 | |||
BC041687.1 | - | 4 | 490 | |||
BC068485.1 | - | 4 | 554 | 30334628 | ||
BQ069026.1 | - | 4 | 579 |