Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031703.2 | NP_001026873.2 | 3 | 309 | ||
XM_011533851.2 | XP_011532153.1 | 3 | 311 | |||
XM_011533852.2 | XP_011532154.1 | 3 | 647 | |||
XM_011533853.2 | XP_011532155.1 | 3 | 288 | |||
XM_017006669.1 | XP_016862158.1 | 3 | 311 | |||
XM_017006670.1 | XP_016862159.1 | 3 | 580 | |||
GenBank mRNA | AK000218.1 | - | 3 | 294 | ||
AK225721.1 | - | 3 | 291 | |||
BC000623.2 | - | 3 | 213 | 3346325 | ||
BC014204.2 | - | 3 | 213 | 4620013 | ||
DQ787088.1 | - | 3 | 190 |