Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031703.2 | NP_001026873.2 | 5 | 632 | ||
XM_005265241.3 | XP_005265298.1 | 4 | 558 | |||
XM_011533851.2 | XP_011532153.1 | 5 | 634 | |||
XM_011533852.2 | XP_011532154.1 | 5 | 970 | |||
XM_011533853.2 | XP_011532155.1 | 5 | 611 | |||
XM_011533854.2 | XP_011532156.1 | 4 | 535 | |||
XM_011533856.2 | XP_011532158.1 | 4 | 590 | |||
XM_011533857.2 | XP_011532159.1 | 4 | 760 | |||
XM_017006669.1 | XP_016862158.1 | 5 | 634 | |||
XM_017006670.1 | XP_016862159.1 | 5 | 903 | |||
GenBank mRNA | AK000218.1 | - | 5 | 617 | ||
AK225721.1 | - | 5 | 614 | |||
AK298202.1 | - | 4 | 567 | |||
BC000623.2 | - | 5 | 536 | 3346325 | ||
BC014204.2 | - | 5 | 536 | 4620013 | ||
DQ787088.1 | - | 5 | 513 |