Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015590.3 | NP_056405.2 | 6, 7 | 490 | ||
NM_182679.2 | NP_872620.1 | 6, 7 | 564 | |||
XM_005245287.3 | XP_005245344.1 | 5, 6 | 486 | |||
GenBank mRNA | AK000256.1 | - | 6, 7 | 557 | ||
AK293026.1 | - | 6, 7 | 558 | |||
BC032436.1 | - | 6, 7 | 543 | |||
BC040147.1 | - | 6, 7 | 547 | |||
CB109133.1 | - | 6, 7 | 473 | |||
DA087471.1 | - | 6, 7 | 564 |