Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083590.1 | NP_001077059.1 | 1 | 240 | ||
NM_017727.4 | NP_060197.4 | 1 | 240 | |||
XM_005264381.3 | XP_005264438.1 | 1 | 213 | |||
XM_005264382.3 | XP_005264439.1 | 1 | 213 | |||
GenBank mRNA | AK074981.1 | - | 1 | 169 | ||
AK097308.1 | - | 1 | 170 | |||
AK296575.1 | - | 1 | 174 | |||
AK297150.1 | - | 1 | 181 | |||
AL833725.1 | - | 1 | 100 | |||
BC068479.1 | - | 1 | 192 | 4814193 | ||
CF994632.1 | - | 1 | 267 |