Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017734.4 | NP_060204.1 | 7 | 1333 | ||
GenBank mRNA | AF262379.1 | - | 7 | 1264 | ||
AJ312214.1 | - | 7 | 1175 | |||
AK000278.1 | - | 7 | 1095 | |||
AK024654.1 | - | 2 | 396 | |||
AL390969.1 | - | 1 | 138 | |||
AL390970.1 | - | 1 | 362 | |||
AL390972.1 | - | 7 | 772 | |||
AL390973.1 | - | 7 | 1059 | |||
BC012570.1 | - | 7 | 1148 | 4245261 | ||
BX648213.1 | - | 4 | 4048 | |||
KU178657.1 | - | 7 | 890 |