Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNBP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024948.2 | NP_001020119.1 | 8 | 813 | ||
NM_001164473.2 | NP_001157945.1 | 8 | 813 | |||
NM_017737.4 | NP_060207.2 | 8 | 813 | |||
XM_011541625.2 | XP_011539927.1 | 8 | 877 | |||
XM_017001533.1 | XP_016857022.1 | 9 | 1008 | |||
GenBank mRNA | AK000282.1 | - | 2 | 65 | ||
AK299760.1 | - | 3 | 199 | |||
AK299943.1 | - | 2 | 109 | |||
AY514449.1 | - | 8 | 662 | |||
BC062477.1 | - | 3 | 264 | |||
BC074891.2 | - | 2 | 43 | |||
BC074892.2 | - | 2 | 43 |