Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161584.1 | NP_001155056.1 | 7 | 1722 | ||
NM_017746.3 | NP_060216.2 | 7 | 1778 | |||
XM_017014848.1 | XP_016870337.1 | 7 | 1267 | |||
GenBank mRNA | AB060968.1 | - | 7 | 1694 | ||
AJ420436.1 | - | 2 | 203 | |||
AK000294.1 | - | 7 | 1694 | |||
AK298916.1 | - | 9 | 1301 | |||
AK302619.1 | - | 7 | 1725 | |||
AY598337.1 | - | 6 | 1563 | |||
BC030652.2 | - | 7 | 1614 | 5268105 |