Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCBTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001268.3 | NP_001259.1 | 13 | 1730 | ||
NM_001286830.1 | NP_001273759.1 | 12 | 1665 | |||
NM_001286831.1 | NP_001273760.1 | 10 | 1470 | |||
NM_001286832.1 | NP_001273761.1 | 8 | 1108 | |||
XM_005266242.2 | XP_005266299.1 | 12 | 1583 | |||
XM_005266244.2 | XP_005266301.1 | 11 | 1518 | |||
XM_006719759.3 | XP_006719822.1 | 12 | 1551 | |||
XM_011534894.2 | XP_011533196.1 | 11 | 1478 | |||
XM_011534895.1 | XP_011533197.1 | 9 | 1199 | |||
XM_017020369.1 | XP_016875858.1 | 14 | 2004 | |||
XM_017020370.1 | XP_016875859.1 | 13 | 1876 | |||
XM_017020371.1 | XP_016875860.1 | 12 | 1749 | |||
XM_017020372.1 | XP_016875861.1 | 11 | 1359 | |||
XM_017020373.1 | XP_016875862.1 | 10 | 1541 | |||
XM_017020378.1 | XP_016875867.1 | 12 | 1536 | |||
XM_017020379.1 | XP_016875868.1 | 11 | 1471 | |||
GenBank mRNA | AB209543.1 | - | 10 | 1436 | ||
AF060219.1 | - | 12 | 1511 | |||
AK023010.1 | - | 12 | 1473 | |||
AK122778.1 | - | 9 | 1385 | |||
AK301516.1 | - | 12 | 1489 | |||
AK304598.1 | - | 8 | 932 | |||
AK310081.1 | - | 11 | 1465 | |||
AK315703.1 | - | 13 | 1650 | |||
BC029052.1 | - | 13 | 1534 | 5194795 |