Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PQLC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040125.1 | NP_001035214.1 | 6, 7 | 1341 | ||
NM_001040126.1 | NP_001035215.1 | 6, 7 | 729 | |||
NM_001287531.1 | NP_001274460.1 | 6, 7 | 698 | |||
NM_017765.2 | NP_060235.2 | 7, 8 | 971 | |||
NR_109848.1 | 7, 8 | 971 | ||||
XM_005245916.2 | XP_005245973.1 | 6, 7 | 754 | |||
XM_006710710.2 | XP_006710773.1 | 7, 8 | 864 | |||
XM_006710711.2 | XP_006710774.1 | 6, 7 | 723 | |||
GenBank mRNA | AK000327.1 | - | 7, 8 | 892 | ||
AK125580.1 | - | 6, 7 | 1259 | |||
AK130171.1 | - | 7, 8 | 889 | |||
AK130175.1 | - | 7, 8 | 891 | |||
AK172745.1 | - | 6, 7 | 616 | |||
BC015324.1 | - | 6, 7 | 634 | 4426038 | ||
KU178659.1 | - | 4, 5 | 424 | |||
KU178660.1 | - | 4, 5 | 393 |