Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136190.1 | NP_001129662.1 | 5 | 1251 | ||
NM_001282878.1 | NP_001269807.1 | 5 | 1082 | |||
NM_017773.3 | NP_060243.2 | 5 | 1522 | |||
XM_006711397.3 | XP_006711460.1 | 6 | 1413 | |||
GenBank mRNA | AK000347.1 | - | 5 | 1084 | ||
AK225331.1 | - | 5 | 1084 | |||
AK301076.1 | - | 5 | 1502 | |||
AK301421.1 | - | 5 | 1224 | |||
AY090784.1 | - | 5 | 1432 | |||
BC069650.1 | - | 4 | 968 | 7262467 | ||
BC089408.1 | - | 5 | 1469 | 3092611 |