Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017774.3 | NP_060244.2 | 16 | 1832 | ||
XM_011514718.1 | XP_011513020.1 | 15 | 1769 | |||
XM_017010987.1 | XP_016866476.1 | 15 | 1725 | |||
GenBank mRNA | AK000349.1 | - | 16 | 1770 | ||
AK024151.1 | - | 3 | 290 | |||
AK128546.1 | - | 14 | 1656 | |||
AK291735.1 | - | 16 | 1812 | |||
BC121020.1 | - | 16 | 1762 | 40117357 | ||
BC121021.1 | - | 16 | 1762 | 40117363 |