Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM208B SIRNA Assays ›
Gene Symbol:
FAM208B
Gene Name:
family with sequence similarity 208 member B
Gene Aliases:
C10orf18, bA318E3.2
Chromosome Location:
Chr.10: 5684759 - 5763740 on Build GRCh38
UniGene ID:
Hs.610717
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29728
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM208B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321783.1
NP_001308712.1
11
1160
NM_001321784.1
NP_001308713.1
11
1206
NM_001321785.1
NP_001308714.1
11
1160
NM_017782.4
NP_060252.4
10
1087
XM_005252474.3
XP_005252531.1
13
7898
XM_005252475.3
XP_005252532.1
14
1559
XM_005252476.4
XP_005252533.1
12
1342
XM_005252480.4
XP_005252537.1
10
1147
XM_005252481.4
XP_005252538.1
8
632
XM_005252482.2
XP_005252539.1
12
992
XM_011519515.2
XP_011517817.1
14
8036
XM_011519516.2
XP_011517818.1
14
8036
XM_011519517.2
XP_011517819.1
14
1956
XM_011519518.2
XP_011517820.1
13
1269
XM_011519519.2
XP_011517821.1
14
8036
XM_011519520.2
XP_011517822.1
11
7733
XM_011519521.2
XP_011517823.1
13
1480
XM_011519522.2
XP_011517824.1
13
1864
XM_011519525.2
XP_011517827.1
11
900
XM_017016361.1
XP_016871850.1
14
8036
XM_017016362.1
XP_016871851.1
12
1763
XM_017016363.1
XP_016871852.1
13
1421
XM_017016364.1
XP_016871853.1
11
1625
XM_017016365.1
XP_016871854.1
12
1287
GenBank mRNA
BX649177.1
-
2
61
CR936780.1
-
10
1088
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-8tsm6:80/100.66.77.47:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline