Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017789.4 | NP_060259.4 | 15 | 2598 | ||
XM_011511378.2 | XP_011509680.1 | 15 | 2706 | |||
XM_011511379.2 | XP_011509681.1 | 15 | 2703 | |||
XM_011511380.1 | XP_011509682.1 | 15 | 3041 | |||
XM_011511381.1 | XP_011509683.1 | 15 | 2675 | |||
XM_011511382.2 | XP_011509684.1 | 15 | 2634 | |||
XM_011511383.1 | XP_011509685.1 | 15 | 2646 | |||
XM_017004393.1 | XP_016859882.1 | 14 | 3128 | |||
XM_017004394.1 | XP_016859883.1 | 13 | 2250 | |||
GenBank mRNA | AB051526.1 | - | 13 | 2828 | ||
AF258577.1 | - | 5 | 1152 | |||
AF370374.1 | - | 2 | 1672 | |||
AK000376.1 | - | 7 | 1538 | |||
AK075388.1 | - | 3 | 1246 | |||
AK126512.1 | - | 3 | 2427 | |||
AY358842.1 | - | 15 | 2571 | |||
BC017476.1 | - | 9 | 1874 | |||
BC062984.1 | - | 7 | 1570 | |||
BC109103.1 | - | 15 | 2527 | 40035252 | ||
BC109104.1 | - | 15 | 2524 | 40035253 |