Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOCAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017794.4 | NP_060264.4 | 41 | 5017 | ||
XM_005251494.3 | XP_005251551.2 | 39 | 5001 | |||
XM_011517945.2 | XP_011516247.1 | 38 | 4605 | |||
XM_017014852.1 | XP_016870341.1 | 40 | 4791 | |||
XM_017014853.1 | XP_016870342.1 | 39 | 4686 | |||
XM_017014854.1 | XP_016870343.1 | 39 | 4686 | |||
XM_017014855.1 | XP_016870344.1 | 40 | 4890 | |||
XM_017014856.1 | XP_016870345.1 | 40 | 4831 | |||
XM_017014857.1 | XP_016870346.1 | 38 | 4896 | |||
XM_017014858.1 | XP_016870347.1 | 38 | 4896 | |||
XM_017014859.1 | XP_016870348.1 | 35 | 4437 | |||
GenBank mRNA | AB058700.1 | - | 27 | 3123 | ||
AK000382.1 | - | 7 | 741 | |||
AK055116.1 | - | 20 | 2325 | |||
AK172818.1 | - | 13 | 1576 | |||
AK225483.1 | - | 7 | 741 | |||
AY139834.1 | - | 41 | 5016 | |||
BC001246.1 | - | 7 | 757 |