Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017798.3 | NP_060268.2 | 4 | 1784 | ||
GenBank mRNA | AB055518.1 | - | 1 | 1125 | ||
AK000398.1 | - | 2 | 945 | |||
AK095795.1 | - | 4 | 1502 | |||
AK300073.1 | - | 2 | 1514 | |||
AL834366.1 | - | 1 | 905 | |||
BC003681.1 | - | 3 | 1458 | |||
BC016920.2 | - | 1 | 1201 | |||
BC025264.1 | - | 1 | 1321 | |||
BC050284.1 | - | 4 | 1784 | 5298438 |