Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017799.3 | NP_060269.3 | 12 | 1534 | ||
XM_005267771.1 | XP_005267828.1 | 9 | 1105 | |||
XM_006720176.1 | XP_006720239.1 | 11 | 1323 | |||
XM_006720178.1 | XP_006720241.1 | 10 | 1208 | |||
XM_011536851.1 | XP_011535153.1 | 12 | 1536 | |||
XM_011536852.1 | XP_011535154.1 | 12 | 1526 | |||
XM_011536853.1 | XP_011535155.1 | 13 | 1704 | |||
XM_017021379.1 | XP_016876868.1 | 12 | 1536 | |||
XM_017021380.1 | XP_016876869.1 | 11 | 1323 | |||
GenBank mRNA | AK000399.1 | - | 9 | 932 | ||
AK023885.1 | - | 4 | 876 | |||
AK056291.1 | - | 12 | 1534 | |||
AK293099.1 | - | 12 | 1501 | |||
AK295989.1 | - | 10 | 1151 | |||
BC008949.2 | - | 11 | 1371 | |||
BC023579.2 | - | 11 | 1371 | |||
BC121163.1 | - | 11 | 1271 | 40121557 | ||
BC121164.1 | - | 12 | 1443 | |||
BX647387.1 | - | 11 | 1134 |