Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017799.3 | NP_060269.3 | 7 | 962 | ||
XM_005267771.1 | XP_005267828.1 | 5 | 648 | |||
XM_006720176.1 | XP_006720239.1 | 6 | 751 | |||
XM_006720178.1 | XP_006720241.1 | 6 | 751 | |||
XM_011536851.1 | XP_011535153.1 | 7 | 964 | |||
XM_011536852.1 | XP_011535154.1 | 7 | 954 | |||
XM_011536853.1 | XP_011535155.1 | 8 | 1132 | |||
XM_017021379.1 | XP_016876868.1 | 7 | 964 | |||
XM_017021380.1 | XP_016876869.1 | 6 | 751 | |||
GenBank mRNA | AK000399.1 | - | 4 | 360 | ||
AK056291.1 | - | 7 | 962 | |||
AK293099.1 | - | 7 | 929 | |||
AK295989.1 | - | 6 | 751 | |||
AK309920.1 | - | 7 | 924 | |||
BC008949.2 | - | 7 | 914 | |||
BC023579.2 | - | 7 | 914 | |||
BC121163.1 | - | 7 | 871 | 40121557 | ||
BC121164.1 | - | 7 | 871 | |||
BX647387.1 | - | 7 | 734 |