Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHTF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039690.3 | NP_001034779.1 | 3 | 242 | ||
NM_001040146.3 | NP_001035236.1 | 3 | 215 | |||
NR_033227.1 | 2 | 184 | ||||
XM_017023370.1 | XP_016878859.1 | 3 | 237 | |||
GenBank mRNA | AF525164.1 | - | 3 | 222 | ||
AK000407.1 | - | 3 | 222 | |||
BC018700.2 | - | 3 | 189 | 3504183 | ||
BC069059.1 | - | 2 | 105 | |||
BP195395.1 | - | 2 | 164 | |||
CB135653.1 | - | 3 | 230 | |||
CB147424.1 | - | 3 | 230 | |||
CN388626.1 | - | 3 | 208 | |||
DA076128.1 | - | 3 | 233 | |||
DA378951.1 | - | 3 | 206 | |||
DB188088.1 | - | 3 | 222 |