Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHTF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039690.3 | NP_001034779.1 | 3 | 268 | ||
NM_001040146.3 | NP_001035236.1 | 3 | 241 | |||
NR_033227.1 | 2 | 210 | ||||
XM_017023370.1 | XP_016878859.1 | 3 | 263 | |||
GenBank mRNA | AF525164.1 | - | 3 | 248 | ||
AK000407.1 | - | 3 | 248 | |||
BC018700.2 | - | 3 | 215 | 3504183 | ||
BC069059.1 | - | 2 | 131 | |||
BP195395.1 | - | 2 | 190 | |||
BP222147.1 | - | 3 | 269 | |||
CB135653.1 | - | 3 | 256 | |||
CB147424.1 | - | 3 | 256 | |||
CN388626.1 | - | 3 | 234 | |||
DA076128.1 | - | 3 | 259 | |||
DA378951.1 | - | 3 | 232 | |||
DB188088.1 | - | 3 | 248 |