Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017805.2 | NP_060275.2 | 11 | 2757 | ||
XM_011527053.2 | XP_011525355.2 | 11 | 2877 | |||
XM_011527054.2 | XP_011525356.2 | 11 | 2877 | |||
XM_011527055.2 | XP_011525357.2 | 8 | 2583 | |||
XM_017026914.1 | XP_016882403.1 | 11 | 2877 | |||
XM_017026915.1 | XP_016882404.1 | 8 | 2583 | |||
GenBank mRNA | AK000408.1 | - | 7 | 872 | ||
AK123179.1 | - | 3 | 1054 | |||
AY378097.1 | - | 11 | 2622 | |||
BC021860.1 | - | 7 | 887 | |||
BC028614.1 | - | 11 | 2629 | 4815880 | ||
BX389914.2 | - | 3 | 256 |