Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145901.1 | NP_001139373.1 | 17 | 1720 | ||
NM_017827.3 | NP_060297.1 | 16 | 1714 | |||
GenBank mRNA | AB029948.1 | - | 16 | 1714 | ||
AK000457.1 | - | 16 | 1581 | |||
AK098522.1 | - | 16 | 1292 | |||
AK293414.1 | - | 17 | 1585 | |||
BC001020.2 | - | 5 | 426 | |||
BC042912.1 | - | 16 | 1568 | 5478759 |