Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM206A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017832.3 | NP_060302.1 | 3 | 252 | ||
XM_011518816.2 | XP_011517118.1 | 3 | 248 | |||
GenBank mRNA | AF153417.1 | - | 3 | 182 | ||
AK000464.1 | - | 3 | 236 | |||
BC015795.1 | - | 3 | 464 | 3910513 |