Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLC41A3 SIRNA Assays ›
Gene Symbol:
SLC41A3
Gene Name:
solute carrier family 41 member 3
Gene Aliases:
SLC41A1-L2
Chromosome Location:
Chr.3: 126006357 - 126101550 on Build GRCh38
UniGene ID:
Hs.573007
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29822
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLC41A3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001008485.1
NP_001008485.1
9, 10
1334
NM_001008486.1
NP_001008486.1
8, 9
1226
NM_001008487.1
NP_001008487.1
8, 9
1253
NM_001164475.1
NP_001157947.1
8, 9
955
NM_017836.3
NP_060306.3
9, 10
1334
XM_005247559.1
XP_005247616.1
9, 10
1212
XM_005247562.1
XP_005247619.1
9, 10
1228
XM_005247563.2
XP_005247620.1
9, 10
1257
XM_005247564.1
XP_005247621.1
9, 10
1212
XM_005247565.2
XP_005247622.1
8, 9
1316
XM_006713681.2
XP_006713744.1
9, 10
1174
XM_011512945.1
XP_011511247.1
8, 9
1361
XM_017006705.1
XP_016862194.1
9, 10
1201
XM_017006706.1
XP_016862195.1
9, 10
1228
XM_017006707.1
XP_016862196.1
9, 10
1251
XM_017006708.1
XP_016862197.1
8, 9
1059
XM_017006709.1
XP_016862198.1
8, 9
1114
GenBank mRNA
AK000480.1
-
8, 9
1057
AK021925.1
-
9, 10
1020
AK022780.1
-
8, 9
1055
AK091671.1
-
10, 11
1138
AK093380.1
-
8, 9
1250
AK094051.1
-
4, 5
1343
AK128054.1
-
4, 5
3368
AK298070.1
-
8, 9
964
AW938624.1
-
2, 3
210
BC009039.2
-
9, 10
1167
3852588
BC009444.1
-
9, 10
1152
BC028241.1
-
8, 9
1150
5243239
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Function(s)
Process(es)
transmembrane transport
cation transmembrane transport
protein binding
cation transmembrane transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-pfczg:80/100.66.78.229:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline