Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COMMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017845.3 | NP_060315.1 | 4 | 486 | ||
XM_017008330.1 | XP_016863819.1 | 4 | 529 | |||
GenBank mRNA | AF182421.1 | - | 4 | 516 | ||
AK000509.1 | - | 4 | 506 | |||
AY542163.1 | - | 4 | 478 | |||
BC008371.1 | - | 4 | 546 | 4051246 | ||
BC015145.1 | - | 4 | 490 | 4052767 | ||
BC019826.1 | - | 4 | 512 | 4995315 |