Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316943.1 | NP_001303872.1 | 1 | 572 | ||
NM_001316944.1 | NP_001303873.1 | 1 | 572 | |||
NM_017851.5 | NP_060321.3 | 1 | 572 | |||
XM_006720589.2 | XP_006720652.1 | 1 | 613 | |||
XM_006720590.2 | XP_006720653.1 | 1 | 613 | |||
XM_006720592.2 | XP_006720655.1 | 1 | 613 | |||
XM_011521742.2 | XP_011520044.1 | 1 | 615 | |||
XM_017022386.1 | XP_016877875.1 | 1 | 613 | |||
XM_017022387.1 | XP_016877876.1 | 1 | 615 | |||
XM_017022388.1 | XP_016877877.1 | 1 | 613 | |||
XM_017022389.1 | XP_016877878.1 | 1 | 613 | |||
GenBank mRNA | AK000516.1 | - | 1 | 572 | ||
AK058010.1 | - | 1 | 156 | |||
BC006389.1 | - | 1 | 418 | 4127509 | ||
BC031074.1 | - | 1 | 221 | 5274010 | ||
DA110379.1 | - | 1 | 435 |