Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNL4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142317.1 | NP_001135789.1 | 4 | 584 | ||
NM_001142318.1 | NP_001135790.1 | 4 | 482 | |||
NM_001324355.1 | NP_001311284.1 | 5 | 733 | |||
NM_017853.2 | NP_060323.1 | 4 | 633 | |||
GenBank mRNA | AK000518.1 | - | 4 | 608 | ||
AK222889.1 | - | 4 | 600 | |||
AY566808.1 | - | 3 | 322 | |||
BC009646.1 | - | 4 | 574 | 3458545 | ||
BM849401.1 | - | 5 | 634 | |||
CD517462.1 | - | 4 | 530 | |||
CR457240.1 | - | 3 | 322 |