Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017867.2 | NP_060337.2 | 8 | 1019 | ||
GenBank mRNA | AK000541.1 | - | 8 | 974 | ||
AK090742.1 | - | 1 | 1469 | |||
AK124036.1 | - | 1 | 2041 | |||
AL136673.1 | - | 8 | 989 | |||
BC010367.1 | - | 8 | 985 | 4334172 |
Gene Symbol: LOC389705 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003920.1 | 5 | 728 |