Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017867.2 | NP_060337.2 | 2 | 174 | ||
XM_017008332.1 | XP_016863821.1 | 2 | 186 | |||
GenBank mRNA | AK000541.1 | - | 2 | 129 | ||
AL136673.1 | - | 2 | 144 | |||
BC010367.1 | - | 2 | 140 | 4334172 | ||
BM905677.1 | - | 2 | 94 | |||
CB141427.1 | - | 2 | 174 | |||
DN990093.1 | - | 2 | 141 |
Gene Symbol: LOC389705 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003920.1 | 2 | 312 | |||
GenBank mRNA | BG719281.1 | - | 2 | 315 |