Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317824.1 | NP_001304753.1 | 4 | 547 | ||
NM_001317825.1 | NP_001304754.1 | 4 | 638 | |||
NM_001317826.1 | NP_001304755.1 | 5 | 610 | |||
NM_017872.4 | NP_060342.2 | 4 | 642 | |||
XM_017009603.1 | XP_016865092.1 | 4 | 577 | |||
GenBank mRNA | AK000553.1 | - | 4 | 618 | ||
AK021663.1 | - | 4 | 593 | |||
AK023355.1 | - | 3 | 256 | |||
AK223119.1 | - | 4 | 611 | |||
AK304590.1 | - | 4 | 518 | |||
AL136669.1 | - | 4 | 610 | |||
AY463216.1 | - | 4 | 716 | |||
BC001523.2 | - | 4 | 588 | 3010614 | ||
BC001852.2 | - | 4 | 588 | 3010614 | ||
BC023521.2 | - | 4 | 608 | 4126840 | ||
CR533503.1 | - | 4 | 590 |