Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000249.3 | NP_000240.1 | 7 | 772 | ||
NM_001167617.1 | NP_001161089.1 | 7 | 567 | |||
NM_001167618.1 | NP_001161090.1 | 7 | 567 | |||
NM_001167619.1 | NP_001161091.1 | 6 | 480 | |||
NM_001258271.1 | NP_001245200.1 | 7 | 772 | |||
NM_001258273.1 | NP_001245202.1 | 6 | 681 | |||
NM_001258274.1 | NP_001245203.1 | 8 | 717 | |||
XM_005265161.1 | XP_005265218.1 | 7 | 662 | |||
XM_005265163.1 | XP_005265220.1 | 7 | 572 | |||
XM_005265164.1 | XP_005265221.1 | 7 | 625 | |||
XM_005265166.1 | XP_005265223.1 | 7 | 580 | |||
XM_011533727.1 | XP_011532029.1 | 7 | 580 | |||
XM_017006449.1 | XP_016861938.1 | 7 | 580 | |||
XM_017006450.1 | XP_016861939.1 | 7 | 625 | |||
XM_017006451.1 | XP_016861940.1 | 7 | 625 | |||
GenBank mRNA | AB209848.1 | - | 7 | 550 | ||
AK222810.1 | - | 7 | 579 | |||
AK295359.1 | - | 7 | 546 | |||
AK298324.1 | - | 7 | 518 | |||
AK298583.1 | - | 7 | 540 | |||
AK302807.1 | - | 9 | 501 | |||
AK311326.1 | - | 7 | 782 | |||
AK311365.1 | - | 7 | 521 | |||
AK312609.1 | - | 7 | 588 | |||
AK316074.1 | - | 6 | 453 | |||
AK316264.1 | - | 6 | 498 | |||
AY517558.1 | - | 7 | 572 | |||
BC006850.1 | - | 7 | 624 | 3451538 | ||
BX648844.1 | - | 8 | 721 | |||
DQ648889.1 | - | 3 | 174 | |||
U07343.1 | - | 7 | 595 | |||
U07418.1 | - | 7 | 615 |