Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085471.1 | NP_001078940.1 | 2 | 569 | ||
NM_005197.3 | NP_005188.2 | 2 | 686 | |||
GenBank mRNA | AK225967.1 | - | 4 | 420 | ||
AK308053.1 | - | 2 | 568 | |||
BC007506.2 | - | 2 | 580 | 3028787 | ||
BP213901.1 | - | 4 | 439 | |||
U68723.1 | - | 2 | 564 |