Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ULK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322500.1 | NP_001309429.1 | 29 | 3354 | ||
NM_001322501.1 | NP_001309430.1 | 28 | 3278 | |||
NM_017886.3 | NP_060356.2 | 29 | 3354 | |||
NR_136342.1 | 27 | 3189 | ||||
XM_011533874.2 | XP_011532176.1 | 29 | 3932 | |||
XM_011533880.2 | XP_011532182.1 | 13 | 1949 | |||
XM_017006711.1 | XP_016862200.1 | 28 | 3848 | |||
XM_017006712.1 | XP_016862201.1 | 29 | 3929 | |||
GenBank mRNA | AK093396.1 | - | 8 | 606 | ||
AK304341.1 | - | 29 | 2985 | |||
AL133104.1 | - | 1 | 26 |